Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hist1h4i | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK011560.1 | - | 1 | 492 | ||
BC019757.1 | - | 1 | 484 | |||
BC052219.1 | - | 1 | 464 | |||
BC092144.1 | - | 1 | 481 | 30612926 | ||
BC125598.1 | - | 1 | 478 | 40130113 | ||
BC125600.1 | - | 1 | 475 | 40130115 |