Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klra17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133203.5 | NP_573466.3 | 4 | 437 | ||
GenBank mRNA | AB033769.1 | - | 4 | 450 | ||
AB190779.1 | - | 3 | 296 | |||
AB190787.1 | - | 3 | 296 | |||
AB190788.1 | - | 3 | 296 | |||
AB190789.1 | - | 3 | 296 | |||
AB193832.1 | - | 3 | 296 | |||
AK080021.1 | - | 4 | 438 | |||
AY642595.1 | - | 3 | 300 | |||
BC132236.1 | - | 4 | 398 | 40130513 |