Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDC42SE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038707.1 | NP_001033796.1 | 5 | 776 | ||
NM_020239.3 | NP_064624.1 | 4 | 640 | |||
XM_017001847.1 | XP_016857336.1 | 5 | 720 | |||
GenBank mRNA | AF187845.2 | - | 4 | 565 | ||
AF286041.1 | - | 3 | 200 | |||
AF286592.1 | - | 3 | 1117 | |||
BC012796.2 | - | 3 | 384 | 3924598 | ||
BC041604.1 | - | 4 | 602 | 5473010 | ||
BX339337.2 | - | 5 | 688 |