Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trappc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024206.2 | NP_001019377.1 | 5 | 519 | ||
GenBank mRNA | BC115818.1 | - | 4 | 488 | 40107010 | |
BC115921.1 | - | 4 | 488 | 40107011 | ||
BY479576.1 | - | 3 | 380 | |||
CK621128.1 | - | 5 | 519 |