Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lonrf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029878.1 | NP_001025049.1 | 11 | 4603 | ||
GenBank mRNA | AK132156.1 | - | 1 | 1406 | ||
BC096025.1 | - | 3 | 3175 | |||
BC139127.1 | - | 11 | 4657 | 8862149 | ||
BC139130.1 | - | 11 | 4654 | 8862152 |