Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2c66 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011707.1 | NP_001011707.1 | 9 | 1590 | ||
GenBank mRNA | AK008512.2 | - | 8 | 1416 | ||
AY227734.1 | - | 9 | 1590 |
Gene Symbol: Cyp2c65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028191.2 | NP_082467.2 | 9 | 1583 | ||
GenBank mRNA | AK008688.1 | - | 9 | 1513 | ||
BC022634.1 | - | 5 | 865 | |||
BC051089.1 | - | 9 | 1567 |