Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278596.1 | NP_001265525.1 | 3 | 344 | ||
NM_001278597.1 | NP_001265526.1 | 3 | 362 | |||
XM_006718988.3 | XP_006719051.1 | 3 | 634 | |||
XM_011520975.2 | XP_011519277.1 | 3 | 2920 | |||
XM_017019672.1 | XP_016875161.1 | 3 | 634 | |||
GenBank mRNA | AB593159.1 | - | 3 | 344 | ||
AJ272204.1 | - | 3 | 362 | |||
FJ384095.1 | - | 3 | 318 |