Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bpifa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291079.1 | NP_001278008.1 | 2 | 358 | ||
NM_028528.3 | NP_082804.1 | 2 | 358 | |||
GenBank mRNA | AK006829.1 | - | 2 | 348 | ||
BC049691.1 | - | 2 | 347 | 6705503 |