Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc10a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009981.2 | NP_001009981.1 | 3 | 462 | ||
NM_001282108.1 | NP_001269037.1 | 3 | 462 | |||
NM_001282109.1 | NP_001269038.1 | 3 | 462 | |||
NM_029736.2 | NP_084012.1 | 3 | 462 | |||
GenBank mRNA | AK010834.1 | - | 3 | 412 | ||
AK040517.1 | - | 3 | 341 | |||
AK079901.1 | - | 3 | 439 | |||
AK152752.1 | - | 3 | 423 | |||
AK152849.1 | - | 3 | 423 | |||
AK160402.1 | - | 3 | 446 | |||
AK165983.1 | - | 3 | 399 | |||
AY825926.1 | - | 3 | 215 | |||
AY825927.1 | - | 2 | 132 | |||
AY825928.1 | - | 3 | 215 | |||
BC116666.1 | - | 3 | 328 | 40107077 | ||
BC116717.1 | - | 3 | 328 | 40107078 |