Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ocel1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293800.1 | NP_001280729.1 | 3 | 771 | ||
NM_029865.2 | NP_084141.2 | 4 | 937 | |||
GenBank mRNA | AK020528.1 | - | 4 | 651 | ||
AK163762.1 | - | 772 | ||||
BB766991.1 | - | 3 | 368 | |||
BC019407.1 | - | 4 | 893 | 5151818 | ||
BY542600.1 | - | 3 | 209 |