Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tas2r116 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053212.1 | NP_444442.1 | 1 | 505 | ||
GenBank mRNA | AF247734.1 | - | 1 | 412 |