Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crebbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025432.1 | NP_001020603.1 | 31 | 6105 | ||
GenBank mRNA | S66385.1 | - | 31 | 5924 |