Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atad5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029856.2 | NP_001025027.1 | 13, 14 | 3781 | ||
GenBank mRNA | AY557610.1 | - | 13, 14 | 3772 | ||
BC046463.1 | - | 9, 10 | 1078 | |||
BC141180.1 | - | 12, 13 | 3277 | 9055792 |