Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr2c2ap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025586.2 | NP_001020757.1 | 5 | 982 | ||
NM_001025587.2 | NP_001020758.1 | 5 | 980 | |||
GenBank mRNA | AK010864.1 | - | 4 | 871 | ||
AK160346.1 | - | 5 | 963 | |||
AK168268.1 | - | 3 | 1298 | |||
AK217197.1 | - | 1 | 38 | |||
BC092288.1 | - | 5 | 924 | 5003230 |