Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmsb15a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030106.2 | NP_084382.1 | 3 | 262 | ||
GenBank mRNA | AK018998.1 | - | 3 | 264 | ||
AK028086.1 | - | 3 | 226 |