Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srbd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030133.3 | NP_084409.3 | 18 | 2280 | ||
GenBank mRNA | AK144490.1 | - | 2 | 129 | ||
BC038140.1 | - | 2 | 224 | |||
BC100359.1 | - | 11 | 1144 | |||
BC147429.1 | - | 2 | 157 | 9056482 | ||
BC147430.1 | - | 2 | 157 | 9056479 | ||
BC172066.1 | - | 3 | 264 | 9054352 |