Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rapgef6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252494.1 | NP_001239423.1 | 29 | 5232 | ||
NM_001252496.1 | NP_001239425.1 | 29 | 4953 | |||
NM_175258.4 | NP_780467.2 | 28 | 5217 | |||
GenBank mRNA | AK143331.1 | - | 16 | 3604 | ||
AK171771.1 | - | 2 | 784 | |||
AK220459.1 | - | 17 | 3145 | |||
BC019702.1 | - | 7 | 1794 | |||
BC025553.1 | - | 11 | 2711 | |||
BC059847.1 | - | 13 | 3509 | |||
BC092525.1 | - | 9 | 1967 | |||
BC141182.1 | - | 29 | 5131 | 9055794 | ||
BC145503.1 | - | 29 | 4853 |