Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037321.1 | NP_001032398.1 | 2 | 1649 | ||
GenBank mRNA | AK036385.1 | - | 3 | 1407 | ||
AK036684.1 | - | 3 | 1403 | |||
AK220373.1 | - | 1 | 1283 | |||
BC107406.1 | - | 2 | 1649 | 40058923 |