Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fancd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033244.3 | NP_001028416.2 | 12, 13 | 1082 | ||
GenBank mRNA | AK142384.1 | - | 12, 13 | 1080 | ||
AK155602.1 | - | 12, 13 | 1061 | |||
AK166411.1 | - | 12, 13 | 1052 | |||
BC139004.1 | - | 12, 13 | 1080 | 8862026 | ||
CV559626.1 | - | 4, 5 | 419 |