Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc27a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011988.2 | NP_036118.2 | 10 | 2137 | ||
GenBank mRNA | AJ577573.1 | - | 10 | 2137 | ||
AK037042.1 | - | 10 | 1897 | |||
AK076014.1 | - | 10 | 1869 | |||
AK131156.1 | - | 4 | 3400 | |||
AK135190.1 | - | 10 | 2114 | |||
BC060052.1 | - | 8 | 2362 | |||
BC141134.1 | - | 10 | 2102 | 9055746 | ||
CJ309971.1 | - | 2 | 273 |