Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030294.1 | NP_001025465.1 | 7 | 1088 | ||
GenBank mRNA | AK133536.1 | - | 5 | 991 | ||
BC141127.1 | - | 5 | 1040 | 9055739 | ||
BC145436.1 | - | 5 | 1040 | 9053994 |