Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Specc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029936.3 | NP_001025107.1 | 13 | 5886 | ||
NM_001281818.1 | NP_001268747.1 | 15 | 6103 | |||
GenBank mRNA | AK012725.1 | - | 1 | 948 | ||
AK220465.1 | - | 13 | 5813 | |||
BC030438.1 | - | 1 | 220 |