Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufs5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030274.1 | NP_001025445.1 | 2 | 186 | ||
GenBank mRNA | AK148505.1 | - | 2 | 185 | ||
AK152845.1 | - | 2 | 181 | |||
AK186159.1 | - | 1 | 65 | |||
BC081434.1 | - | 2 | 130 | 6412214 | ||
BU534760.1 | - | 2 | 189 |