Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Setd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035123.3 | NP_001030295.1 | 3 | 569 | ||
GenBank mRNA | AK010304.1 | - | 3 | 570 | ||
AK075597.1 | - | 3 | 572 | |||
AK167753.1 | - | 3 | 551 | |||
AK181998.1 | - | 1 | 56 | |||
AK215299.1 | - | 1 | 56 | |||
AK215534.1 | - | 1 | 73 | |||
BC139198.1 | - | 3 | 601 | 8862220 | ||
BC139199.1 | - | 3 | 601 | 8862221 |