Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Diras2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024474.2 | NP_001019645.1 | 2 | 1731 | ||
GenBank mRNA | AK136096.1 | - | 2 | 1732 | ||
AK136101.1 | - | 2 | 1732 | |||
BC086799.1 | - | 2 | 1549 | 6833911 |