Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hddc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027168.2 | NP_081444.1 | 2 | 129 | ||
GenBank mRNA | AK009957.1 | - | 2 | 70 | ||
AK161186.1 | - | 1 | 1160 | |||
BC104360.1 | - | 2 | 129 | 40049707 | ||
BC104361.1 | - | 2 | 129 | 40049709 |