Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2310003H01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027980.2 | NP_082256.2 | 5 | 2064 | ||
GenBank mRNA | AK082976.1 | - | 5 | 2067 | ||
AK147900.1 | - | 2155 | ||||
AK148234.1 | - | 5 | 2051 | |||
AK165631.1 | - | 4 | 1935 | |||
AK193773.1 | - | 1 | 63 | |||
BC034203.1 | - | 5 | 1935 | 4986832 | ||
BC048376.1 | - | 1 | 403 |