Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2700094K13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033166.2 | NP_001028338.1 | 2 | 304 | ||
NM_001037279.1 | NP_001032356.1 | 2 | 304 | |||
GenBank mRNA | AK012609.1 | - | 2 | 282 | ||
AK142622.1 | - | 2 | 294 | |||
AK167638.1 | - | 2 | 276 | |||
AK171972.1 | - | 2 | 295 | |||
BC056177.1 | - | 2 | 297 | 5683334 | ||
BY239096.1 | - | 2 | 366 | |||
CA318611.1 | - | 2 | 201 |