Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Macrod2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028387.1 | NP_082663.1 | 4 | 2826 | ||
GenBank mRNA | AK029184.1 | - | 4 | 2806 |