Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr75 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028599.2 | NP_082875.1 | 11 | 1118 | ||
GenBank mRNA | AK010772.1 | - | 11 | 1118 | ||
AK152412.1 | - | 11 | 1119 | |||
AK163221.1 | - | 11 | 1092 | |||
AK168032.1 | - | 11 | 1112 | |||
AK169932.1 | - | 11 | 1116 | |||
BC085177.1 | - | 11 | 1148 | |||
BC117880.2 | - | 11 | 1148 | 40096010 | ||
BC117881.1 | - | 11 | 1145 | 40096014 |