Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013026.2 | NP_001013044.2 | 14 | 2339 | ||
GenBank mRNA | AK014607.1 | - | 14 | 2327 | ||
AK078759.1 | - | 14 | 2339 | |||
AK087352.1 | - | 14 | 2339 | |||
BC087733.1 | - | 14 | 2346 | 30434981 | ||
BC096625.1 | - | 14 | 2337 | 30109731 |