Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDHC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035511.1 | NP_001030588.1 | 5 | 1310 | ||
NM_001035512.1 | NP_001030589.1 | 5 | 1372 | |||
NM_001035513.1 | NP_001030590.1 | 4 | 1315 | |||
NM_001278172.1 | NP_001265101.1 | 4 | 1208 | |||
NM_003001.3 | NP_002992.1 | 6 | 1474 | |||
NR_103459.1 | 7 | 1501 |