Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TRO SIRNA Assays ›
Gene Symbol:
TRO
Gene Name:
trophinin
Gene Aliases:
MAGE-d3, MAGED3
Chromosome Location:
Chr.X: 54920563 - 54931436 on Build GRCh38
UniGene ID:
Hs.633653
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
284698
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TRO
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001039705.2
NP_001034794.1
13
4733
NM_001271183.1
NP_001258112.1
11
3453
NM_001271184.1
NP_001258113.1
12
3353
NM_016157.3
NP_057241.2
13
2412
NM_177556.2
NP_808224.1
13
2397
NM_177557.2
NP_808225.1
12
1206
NR_073148.1
14
2688
NR_073149.1
13
1497
XM_006724600.2
XP_006724663.1
13
4499
XM_011530808.1
XP_011529110.1
13
4544
XM_011530809.1
XP_011529111.1
13
4495
XM_011530811.2
XP_011529113.1
13
4881
XM_011530812.1
XP_011529114.1
13
4482
XM_011530813.1
XP_011529115.1
13
4532
XM_011530814.1
XP_011529116.1
13
4426
XM_017029767.1
XP_016885256.1
13
4896
XM_017029768.1
XP_016885257.1
13
4513
XM_017029769.1
XP_016885258.1
13
2174
XM_017029770.1
XP_016885259.1
13
2223
XM_017029771.1
XP_016885260.1
13
2551
XM_017029772.1
XP_016885261.1
13
2192
XM_017029773.1
XP_016885262.1
13
2103
GenBank mRNA
AB029037.2
-
13
4454
AF349719.1
-
14
2435
AF349720.1
-
13
2144
AF349721.1
-
13
1243
AF349722.1
-
12
953
AK001300.1
-
11
1842
AK027489.1
-
13
2151
AK027861.1
-
9
3517
AK225841.1
-
12
3326
AK296329.1
-
11
3192
AK299568.1
-
4110
BC026914.1
-
14
2418
BM693634.1
-
3
430
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
homophilic cell adhesion via plasma membrane adhesion molecules
embryo implantation
negative regulation of cell growth
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0