Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STK19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004197.1 | NP_004188.1 | 2 | 403 | ||
NM_032454.1 | NP_115830.1 | 2 | 403 | |||
NR_026717.1 | 2 | 588 | ||||
GenBank mRNA | AK302111.1 | - | 2 | 602 | ||
AK303438.1 | - | 2 | 441 | |||
BC110347.1 | - | 2 | 588 | 5165123 | ||
BC121128.1 | - | 1 | 422 | 40120738 | ||
BC121129.1 | - | 1 | 421 | 40120740 | ||
BC143833.1 | - | 1 | 458 | 9052348 | ||
BC143835.1 | - | 1 | 458 | |||
L26260.1 | - | 2 | 403 |