Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KAZN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017999.2 | NP_001017999.1 | 2 | 325 | ||
NM_001018000.3 | NP_001018000.1 | 2 | 514 | |||
NM_001018001.2 | NP_001018001.1 | 2 | 315 | |||
NM_015209.2 | NP_056024.1 | 2 | 577 | |||
NM_201628.2 | NP_963922.2 | 2 | 577 | |||
XM_005245795.4 | XP_005245852.1 | 4 | 832 | |||
XM_011541074.2 | XP_011539376.1 | 4 | 839 | |||
XM_011541075.2 | XP_011539377.1 | 2 | 495 | |||
XM_011541076.2 | XP_011539378.1 | 2 | 640 | |||
XM_011541077.2 | XP_011539379.1 | 2 | 1619 | |||
XM_011541080.2 | XP_011539382.1 | 4 | 832 | |||
XM_017000768.1 | XP_016856257.1 | 4 | 832 | |||
XM_017000769.1 | XP_016856258.1 | 4 | 832 | |||
XM_017000770.1 | XP_016856259.1 | 4 | 832 | |||
XM_017000771.1 | XP_016856260.1 | 3 | 500 | |||
XM_017000772.1 | XP_016856261.1 | 2 | 179 | |||
GenBank mRNA | AY505119.1 | - | 3 | 505 | ||
AY505120.1 | - | 2 | 514 | |||
AY505121.1 | - | 2 | 315 | |||
AY505122.1 | - | 2 | 325 | |||
BC035501.2 | - | 2 | 513 | 4556194 | ||
BI916330.1 | - | 2 | 577 |