Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCRL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159488.1 | NP_001152960.1 | 9 | 2075 | ||
NM_030764.3 | NP_110391.2 | 12 | 2256 | |||
NR_125358.1 | 8 | 1338 | ||||
XM_011509974.2 | XP_011508276.1 | 12 | 2617 | |||
XM_011509975.2 | XP_011508277.1 | 11 | 2553 | |||
XM_011509976.2 | XP_011508278.1 | 11 | 2526 | |||
XM_017002316.1 | XP_016857805.1 | 11 | 2562 | |||
XM_017002317.1 | XP_016857806.1 | 11 | 2365 | |||
XM_017002318.1 | XP_016857807.1 | 10 | 2285 | |||
XM_017002319.1 | XP_016857808.1 | 10 | 2277 | |||
GenBank mRNA | AF459633.1 | - | 12 | 2244 | ||
AL833194.1 | - | 7 | 4115 | |||
AL833361.1 | - | 2 | 2776 | |||
AY043465.1 | - | 12 | 2243 | |||
BC069185.1 | - | 8 | 1342 |