Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPD52L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001874.2 | NP_001001874.1 | 1 | 348 | ||
NM_001001875.3 | NP_001001875.1 | 1 | 348 | |||
NM_033516.5 | NP_277051.3 | 1 | 348 | |||
XM_017015280.1 | XP_016870769.1 | 1 | 336 | |||
GenBank mRNA | AK057479.1 | - | 1 | 322 | ||
AY032877.1 | - | 1 | 326 | |||
BC017589.2 | - | 1 | 322 | 4828082 | ||
BC033792.1 | - | 1 | 293 | 5163651 | ||
BC042113.1 | - | 1 | 307 | 4838253 | ||
BP369721.1 | - | 1 | 348 |