Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLDN23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194284.2 | NP_919260.2 | 1 | 1577 | ||
GenBank mRNA | AK123547.1 | - | 2 | 1202 |