Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAMD13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010971.2 | NP_001010971.1 | 4 | 675 | ||
NM_001134663.1 | NP_001128135.1 | 4 | 669 | |||
NM_001134664.1 | NP_001128136.1 | 4 | 513 | |||
XM_017000377.1 | XP_016855866.1 | 4 | 1343 | |||
GenBank mRNA | AK056937.1 | - | 4 | 676 | ||
AY604569.1 | - | 2 | 872 | |||
AY604570.1 | - | 2 | 569 | |||
BC037973.1 | - | 4 | 513 | |||
DB459148.2 | - | 4 | 671 |