Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFNL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172138.1 | NP_742150.1 | 6 | 672 | ||
GenBank mRNA | AY129148.1 | - | 6 | 672 | ||
DQ126337.1 | - | 6 | 672 |