Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CORO1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018070.2 | NP_001018080.1 | 7 | 840 | ||
NM_020441.2 | NP_065174.1 | 6 | 818 | |||
GenBank mRNA | AK000860.1 | - | 6 | 801 | ||
AK295782.1 | - | 6 | 778 | |||
AK315399.1 | - | 6 | 773 | |||
AL162072.1 | - | 1 | 91 | |||
BC006449.1 | - | 6 | 768 | 3958930 |