Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB8OS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308135.1 | NP_001295064.1 | 6 | 557 | ||
NM_001308136.1 | NP_001295065.1 | 6 | 525 | |||
NM_001308137.1 | NP_001295066.1 | 5 | 504 | |||
NM_001308138.1 | NP_001295067.1 | 6 | 622 | |||
NM_001308139.1 | NP_001295068.1 | 7 | 616 | |||
NM_001308140.1 | NP_001295069.1 | 7 | 609 | |||
NM_001308141.1 | NP_001295070.1 | 7 | 582 | |||
NM_178547.3 | NP_848642.1 | 7 | 578 | |||
XM_011541328.1 | XP_011539630.1 | 7 | 603 | |||
XM_011541329.2 | XP_011539631.1 | 7 | 503 | |||
XM_011541330.2 | XP_011539632.1 | 6 | 989 | |||
XM_011541334.2 | XP_011539636.1 | 6 | 948 | |||
XM_011541335.2 | XP_011539637.1 | 7 | 555 | |||
XM_011541337.2 | XP_011539639.1 | 7 | 590 | |||
XM_011541338.2 | XP_011539640.1 | 6 | 949 | |||
XM_017001134.1 | XP_016856623.1 | 7 | 589 | |||
XM_017001136.1 | XP_016856625.1 | 5 | 800 | |||
XM_017001137.1 | XP_016856626.1 | 7 | 437 | |||
XM_017001138.1 | XP_016856627.1 | 6 | 475 | |||
GenBank mRNA | AK289480.1 | - | 7 | 920 | ||
AY099494.1 | - | 7 | 478 | |||
AY151084.1 | - | 7 | 572 | |||
BC058843.1 | - | 6 | 495 | 6745279 | ||
BE780045.1 | - | 6 | 517 | |||
BM465977.1 | - | 2 | 210 | |||
CN344731.1 | - | 6 | 465 |