Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCRP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037566.1 | 5 | 736 | |||
GenBank mRNA | AK124943.1 | - | 5 | 736 |
Gene Symbol: LOC102724751 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017030164.1 | XP_016885653.1 | 2 | 269 | ||
XM_017030165.1 | XP_016885654.1 | 2 | 269 |