Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDRG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130487.1 | NP_001123959.1 | 3, 4 | 526 | ||
NM_020465.3 | NP_065198.1 | 3, 4 | 301 | |||
NM_022910.3 | NP_075061.1 | 3, 4 | 398 | |||
XM_006721257.3 | XP_006721320.1 | 3, 4 | 373 | |||
XM_006721259.3 | XP_006721322.1 | 3, 4 | 342 | |||
XM_006721260.3 | XP_006721323.1 | 3, 4 | 280 | |||
XM_011523293.2 | XP_011521595.1 | 3, 4 | 446 | |||
XM_011523294.2 | XP_011521596.1 | 4, 5 | 267 | |||
XM_017023589.1 | XP_016879078.1 | 3, 4 | 342 | |||
XM_017023590.1 | XP_016879079.1 | 4, 5 | 267 | |||
GenBank mRNA | AB021172.1 | - | 3, 4 | 201 | ||
AB044946.1 | - | 2, 3 | 148 | |||
AK126574.1 | - | 3, 4 | 298 | |||
CF552009.1 | - | 3, 4 | 244 | |||
CR936733.1 | - | 3, 4 | 212 | |||
DA069318.1 | - | 3, 4 | 359 | |||
DA242460.1 | - | 3, 4 | 301 |