Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031692.2 | NP_001026862.1 | 2 | 640 | ||
NM_005824.2 | NP_005815.2 | 2 | 640 | |||
XM_005250108.1 | XP_005250165.1 | 3 | 1056 | |||
GenBank mRNA | AK225701.1 | - | 2 | 537 | ||
AK226021.1 | - | 2 | 546 | |||
AK290305.1 | - | 2 | 531 | |||
AY358889.1 | - | 2 | 476 | |||
BC027903.1 | - | 2 | 518 | 5229061 | ||
U32907.1 | - | 2 | 532 |