Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRIP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020645.2 | NP_065696.1 | 1, 2 | 290 | ||
GenBank mRNA | AK024750.1 | - | 1, 2 | 243 | ||
AK098694.1 | - | 1, 2 | 243 | |||
AK296484.1 | - | 1, 2 | 242 | |||
BC008835.2 | - | 1, 2 | 226 | 4304488 | ||
DA249943.1 | - | 1, 2 | 291 |