Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRTM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304389.1 | NP_001291318.1 | 2 | 465 | ||
NM_020678.3 | NP_065729.1 | 2 | 474 | |||
GenBank mRNA | AF225421.1 | - | 2 | 445 | ||
BC040732.1 | - | 2 | 465 | 5223743 | ||
BC074741.2 | - | 2 | 335 | 30915199 | ||
BM716618.1 | - | 2 | 474 |