Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADGRG6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001032394.2 | NP_001027566.1 | 17 | 2781 | ||
NM_001032395.2 | NP_001027567.1 | 17 | 2781 | |||
NM_020455.5 | NP_065188.4 | 18 | 2865 | |||
NM_198569.2 | NP_940971.1 | 18 | 2865 | |||
XM_005267061.3 | XP_005267118.1 | 18 | 2766 | |||
XM_006715516.2 | XP_006715579.1 | 18 | 2766 | |||
XM_006715517.2 | XP_006715580.1 | 18 | 2566 | |||
XM_006715518.2 | XP_006715581.1 | 17 | 2682 | |||
XM_011535964.1 | XP_011534266.1 | 18 | 2569 | |||
XM_017011085.1 | XP_016866574.1 | 18 | 2766 | |||
GenBank mRNA | AB183546.1 | - | 18 | 2855 | ||
AB183547.1 | - | 17 | 2771 | |||
AB183548.1 | - | 18 | 2855 | |||
AB183549.1 | - | 17 | 2771 | |||
AK027843.1 | - | 6 | 519 | |||
AY181244.1 | - | 327 | ||||
BC075798.1 | - | 17 | 2841 | 30333185 | ||
BX640873.1 | - | 17 | 2387 | |||
BX640971.1 | - | 18 | 2791 | |||
BX648315.1 | - | 17 | 2494 |
Set Membership: |
G-protein coupled receptor |