Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MIF4GD SIRNA Assays ›
Gene Symbol:
MIF4GD
Gene Name:
MIF4G domain containing
Gene Aliases:
AD023, MIFD, SLIP1
Chromosome Location:
Chr.17: 75266228 - 75271292 on Build GRCh38
UniGene ID:
Hs.325631
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
28654
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MIF4GD
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242498.1
NP_001229427.1
4
409
NM_001242500.1
NP_001229429.1
3
311
NM_001242501.1
NP_001229430.1
2
209
NM_020679.3
NP_065730.2
4
388
XM_005257530.1
XP_005257587.1
4
530
XM_005257532.4
XP_005257589.1
3
286
XM_005257533.1
XP_005257590.1
3
385
XM_011525051.2
XP_011523353.1
4
625
XM_011525052.2
XP_011523354.1
4
604
XM_011525053.2
XP_011523355.1
3
502
XM_011525054.2
XP_011523356.1
4
625
XM_011525055.2
XP_011523357.1
3
502
XM_011525057.2
XP_011523359.1
2
205
XM_017024888.1
XP_016880377.1
4
625
XM_017024889.1
XP_016880378.1
4
409
XM_017024890.1
XP_016880379.1
4
435
XM_017024891.1
XP_016880380.1
4
347
XM_017024892.1
XP_016880381.1
3
502
XM_017024893.1
XP_016880382.1
3
781
XM_017024894.1
XP_016880383.1
3
416
XM_017024895.1
XP_016880384.1
4
388
GenBank mRNA
AF225422.1
-
2
405
AK300711.1
-
4
348
BC033759.1
-
3
287
5215495
BI752882.1
-
4
406
BU190758.1
-
3
310
BX404175.2
-
4
331
CA488406.1
-
2
209
KU178731.1
-
3
251
KU178732.1
-
3
272
KU178733.1
-
2
149
KU178734.1
-
2
149
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
regulation of translation
RNA binding
protein binding
protein C-terminus binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0