Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snurf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270712.1 | NP_001257641.1 | 1, 2 | 103 | ||
NM_130738.2 | NP_570094.1 | 1, 2 | 103 | |||
GenBank mRNA | AF101041.1 | - | 1, 2 | 63 |
Gene Symbol: LOC100362965 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002728491.3 | XP_002728537.1 | 1 | 61 |